The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172550.4 | NP_766138.3 | 4 - 5 | 769 | 63 |
GenBank mRNA | AK037727.1 | - | 4 - 5 | 496 | 63 |
AK136543.1 | - | 4 - 5 | 520 | 63 | |
BC057308.1 | - | 4 - 5 | 769 | 63 |