Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tns4
Entrez Gene ID
217169
Gene Name
tensin 4
Gene Aliases
9930017A07Rik, AA589547, AU016405, Cten
Chromosome Location
Chr.11: 99065678 - 99089306 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172564.3 NP_766152.2 4 - 5 1391 63
GenBank mRNA AK036834.1 - 4 - 5 1388 63
BC055820.1 - 4 - 5 1373 63

View Full Product Details