The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172565.2 | NP_766153.1 | 1 - 2 | 601 | 131 |
GenBank mRNA | AK029101.1 | - | 1 - 2 | 602 | 131 |
AK169910.1 | - | 2 - 3 | 499 | 130 | |
BC111873.1 | - | 1 - 2 | 597 | 131 |