Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem63c
Entrez Gene ID
217733
Gene Name
transmembrane protein 63c
Gene Aliases
4932420N09, 9330187M14Rik
Chromosome Location
Chr.12: 87021344 - 87091824 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172583.2 NP_766171.1 12 - 13 1107 104
GenBank mRNA AK030062.1 - 12 - 13 1099 104
AK034410.1 - 11 - 12 1030 104
AK043536.1 - 11 - 12 1028 104
BC056936.1 - 12 - 13 1107 104
BC057088.1 - 12 - 13 1107 104

View Full Product Details