Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem44
Entrez Gene ID
224090
Gene Name
transmembrane protein 44
Gene Aliases
1700007N03Rik, 9330161C17Rik, B230220N21
Chromosome Location
Chr.16: 30511855 - 30550834 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK045675.1 - 11 - 12 1369 99
BC080307.1 - 11 - 12 1202 99

View Full Product Details