Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oscp1
Entrez Gene ID
230751
Gene Name
organic solute carrier partner 1
Gene Aliases
1810007P19Rik, 5730415O04, 6030436A01Rik
Chromosome Location
Chr.4: 126057287 - 126089334 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172701.2 NP_766289.2 6 - 7 869 67
GenBank mRNA AB154826.1 - 6 - 7 822 67
AB286960.1 - 7 - 8 870 67
AK077467.1 - 6 - 7 835 67
AK077909.1 - 6 - 7 790 67
BC045150.1 - 6 - 7 815 67

View Full Product Details