The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168290.1 | NP_001161762.1 | 8 - 9 | 3395 | 76 |
NM_172755.3 | NP_766343.3 | 8 - 9 | 3395 | 76 | |
GenBank mRNA | AF518875.1 | - | 8 - 9 | 3388 | 76 |
AK042293.1 | - | 8 - 9 | 3396 | 76 | |
BC023276.1 | - | 4 - 5 | 1162 | 76 | |
BC042763.1 | - | 6 - 7 | 1743 | 76 | |
BC047383.1 | - | - | 482 | 76 | |
BC057305.1 | - | 8 - 9 | 3048 | 76 |