Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankle1
Entrez Gene ID
234396
Gene Name
ankyrin repeat and LEM domain containing 1
Gene Aliases
8430438L13Rik, Ankrd41, C430003I23, Lem3
Chromosome Location
Chr.8: 71406005 - 71410543 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172756.2 NP_766344.2 2 - 3 242 96
GenBank mRNA AK049395.1 - 2 - 3 233 96
AK078830.1 - 2 - 3 233 96
AK080768.1 - 2 - 3 238 96
AK083552.1 - 2 - 3 225 96
AK087444.1 - 2 - 3 238 96
BC094316.1 - 2 - 3 199 96
CJ046325.1 - 2 - 3 243 96
EU184014.1 - 2 - 3 218 96

View Full Product Details