Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btnl9
Entrez Gene ID
237754
Gene Name
butyrophilin-like 9
Gene Aliases
B430208I01, Btn3, D330012D11Rik
Chromosome Location
Chr.11: 49168325 - 49187089 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172793.2 NP_766381.1 4 - 5 856 82
GenBank mRNA AK080918.1 - 4 - 5 840 82
AK084528.1 - 4 - 5 857 82
BC025540.1 - 4 - 5 834 82
BC052556.1 - 3 - 4 769 82
BC151155.1 - 4 - 5 813 82

View Full Product Details