The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172835.3 | NP_766423.3 | 1 - 2 | 160 | 56 |
GenBank mRNA | AK044418.1 | - | 1 - 2 | 107 | 56 |
BC028931.1 | - | 1 - 2 | 118 | 56 | |
BY128359.1 | - | 1 - 2 | 160 | 56 |