The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172835.3 | NP_766423.3 | 2 - 3 | 304 | 76 |
GenBank mRNA | AK044418.1 | - | 2 - 3 | 251 | 76 |
AK135575.1 | - | 2 - 3 | 253 | 76 | |
BC028931.1 | - | 2 - 3 | 262 | 76 |