Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppp6r1
Entrez Gene ID
243819
Gene Name
protein phosphatase 6, regulatory subunit 1
Gene Aliases
2010309P17Rik, AI836219, B430201G11Rik, D030074N20Rik, Pp6r1, Saps1, mKIAA1115
Chromosome Location
Chr.7: 4631495 - 4658950 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172894.2 NP_766482.2 1 - 2 470 87
GenBank mRNA AK080891.1 - 1 - 2 87
AK122450.1 - 1 - 2 470 87
BC053076.1 - 1 - 2 470 87

View Full Product Details