The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173019.5 | NP_766607.3 | 12 - 13 | 1394 | 64 |
GenBank mRNA | AK048999.1 | - | 4 - 5 | 508 | 64 |
AK155172.1 | - | 12 - 13 | 1332 | 64 | |
AY646092.2 | - | 12 - 13 | 1280 | 64 | |
AY707862.1 | - | 12 - 13 | 1297 | 64 | |
AY756065.1 | - | 12 - 13 | 1387 | 64 | |
AY756066.1 | - | 12 - 13 | 1387 | 64 | |
AY756067.1 | - | 12 - 13 | 1258 | 64 | |
BC057594.1 | - | 12 - 13 | 1346 | 64 | |
DQ350638.1 | - | 13 - 14 | 1366 | 64 | |
EU571215.1 | - | 12 - 13 | 1378 | 64 | |
EU571216.1 | - | 14 - 15 | 1553 | 64 | |
EU571217.1 | - | 12 - 13 | 1521 | 64 |