Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tstd2
Entrez Gene ID
272027
Gene Name
thiosulfate sulfurtransferase (rhodanese)-like domain containing 2
Gene Aliases
3010020C06
Chromosome Location
Chr.4: 46114746 - 46138581 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173033.3 NP_766621.3 7 - 8 1090 68
GenBank mRNA AK028258.1 - 7 - 8 1033 68
AK155456.1 - 7 - 8 1037 68
BC057893.1 - 7 - 8 1025 68
CD549731.1 - 5 - 6 495 68

View Full Product Details