Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sptlc3
Entrez Gene ID
228677
Gene Name
serine palmitoyltransferase, long chain base subunit 3
Gene Aliases
C130053K05Rik
Chromosome Location
Chr.2: 139493913 - 139637674 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175467.3 NP_780676.1 2 - 3 324 86
GenBank mRNA AK048374.1 - 1 - 2 185 86
AK054240.1 - 1 - 2 411 86
AK078679.1 - 1 - 2 388 86
AK078686.1 - 2 - 3 325 86
BC094496.1 - 1 - 2 170 86

View Full Product Details