Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Coro2b
Entrez Gene ID
235431
Gene Name
coronin, actin binding protein, 2B
Gene Aliases
CLIPINC, E130012P22Rik
Chromosome Location
Chr.9: 62419490 - 62537044 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175484.3 NP_780693.2 9 - 10 1457 69
GenBank mRNA AK047527.1 - 9 - 10 1399 69
AK087414.1 - 9 - 10 1458 69
BC062649.1 - 9 - 10 1189 69

View Full Product Details