Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
A2m
Entrez Gene ID
232345
Gene Name
alpha-2-macroglobulin
Gene Aliases
A2mp
Chromosome Location
Chr.6: 121636166 - 121679238 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175628.3 NP_783327.2 14 - 15 1834 91
GenBank mRNA AY185125.1 - 14 - 15 1783 91
BC023844.1 - 4 - 5 496 91
BC072642.1 - 14 - 15 1774 91

View Full Product Details