The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021887.2 | NP_068687.1 | 3 - 4 | 591 | 64 |
GenBank mRNA | AB049137.1 | - | 3 - 4 | 483 | 64 |
AF254068.1 | - | 3 - 4 | 295 | 64 | |
AF269134.1 | - | 2 - 3 | 153 | 64 | |
AF279436.1 | - | 3 - 4 | 543 | 64 | |
AF477982.1 | - | 2 - 3 | 169 | 64 | |
AF477983.1 | - | 2 - 3 | 169 | 64 | |
AF477984.1 | - | 2 - 3 | 169 | 64 | |
AF477985.1 | - | 2 - 3 | 169 | 64 | |
AF477986.1 | - | 2 - 3 | 169 | 64 | |
AK150824.1 | - | 3 - 4 | 552 | 64 | |
AK171826.1 | - | 3 - 4 | 530 | 64 | |
BC057861.1 | - | 3 - 4 | 540 | 64 |