The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153079.4 | NP_694719.2 | 3 - 4 | 1011 | 63 |
GenBank mRNA | AK036756.1 | - | 3 - 4 | 1012 | 63 |
AY057384.1 | - | 3 - 4 | 907 | 63 | |
BC137611.1 | - | 3 - 4 | 940 | 63 |