The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023738.4 | NP_076227.1 | 18 - 19 | 2361 | 64 |
GenBank mRNA | AF288694.1 | - | 13 - 14 | 1623 | 64 |
AK004894.2 | - | 18 - 19 | 2362 | 64 | |
AK054410.1 | - | 17 - 18 | 2453 | 64 | |
AK089235.1 | - | 17 - 18 | 2254 | 64 | |
AK129009.1 | - | 18 - 19 | 2271 | 64 | |
AK157941.1 | - | 1 - 2 | 444 | 64 | |
AK170421.1 | - | 17 - 18 | 2439 | 64 | |
AK170644.1 | - | 16 - 17 | 2820 | 64 | |
AK172467.1 | - | 17 - 18 | 2699 | 64 | |
BC138843.1 | - | 18 - 19 | 2278 | 64 |