The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243041.1 | NP_001229970.1 | 10 - 11 | 1071 | 61 |
NM_134079.4 | NP_598840.1 | 10 - 11 | 1028 | 61 | |
GenBank mRNA | AK050135.1 | - | 4 - 5 | 331 | 61 |
AK151863.1 | - | 10 - 11 | 1073 | 61 | |
AK160264.1 | - | 10 - 11 | 1025 | 61 | |
AY540996.1 | - | 10 - 11 | 970 | 61 | |
BC009659.1 | - | 10 - 11 | 993 | 61 |