Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crybg3
Entrez Gene ID
224273
Gene Name
beta-gamma crystallin domain containing 3
Gene Aliases
5031404N19, AU022790
Chromosome Location
Chr.16: 59490775 - 59601399 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174848.3 NP_777273.2 3 - 4 1806 94
GenBank mRNA BC043118.1 - 3 - 4 1806 94

View Full Product Details