Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Micall2
Entrez Gene ID
231830
Gene Name
MICAL-like 2
Gene Aliases
A930021H16Rik, JRAB, MICAL-L2, mFLJ00139
Chromosome Location
Chr.5: 139706692 - 139736333 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174850.3 NP_777275.2 11 - 12 2678 103
GenBank mRNA AB182579.1 - 11 - 12 2529 103
AK044547.1 - 11 - 12 2530 103
AK151086.1 - 11 - 12 2677 103
AK164043.1 - 11 - 12 2649 103
AK165558.1 - 11 - 12 2528 103
AK220308.1 - 11 - 12 2455 103
BC043300.1 - 11 - 12 2492 103

View Full Product Details