The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162375.1 | NP_001155847.1 | 2 - 3 | 479 | 79 |
NM_174868.4 | NP_777357.2 | 2 - 3 | 479 | 79 | |
GenBank mRNA | AK045347.1 | - | 2 - 3 | 478 | 79 |
AK083909.1 | - | 1 - 2 | 310 | 79 | |
AK085017.1 | - | 2 - 3 | 479 | 79 | |
AK143366.1 | - | 2 - 3 | 478 | 79 | |
AK166051.1 | - | 2 - 3 | 430 | 79 | |
BC059928.1 | - | 2 - 3 | 445 | 79 | |
BC098205.1 | - | 2 - 3 | 462 | 79 | |
BC138328.1 | - | 2 - 3 | 426 | 79 | |
BC138329.1 | - | 2 - 3 | 426 | 79 |