Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsr2
Entrez Gene ID
69499
Gene Name
TSR2 20S rRNA accumulation
Gene Aliases
2310007F12Rik
Chromosome Location
Chr.X: 151087094 - 151096543 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175146.4 NP_780355.2 4 - 5 472 75
NR_028392.1 3 - 4 381 75
GenBank mRNA AK044504.1 - 4 - 5 448 75
BC025008.1 - 4 - 5 435 75
BC037444.1 - 3 - 4 336 75
BC049668.1 - 4 - 5 454 75
BG083382.2 - 3 - 4 345 75

View Full Product Details