Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl5
Entrez Gene ID
71778
Gene Name
kelch-like 5
Gene Aliases
1300013C10Rik
Chromosome Location
Chr.5: 65107527 - 65168186 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175174.4 NP_780383.2 1 - 2 546 86
GenBank mRNA AK090246.1 - 1 - 2 106 86
BC057606.1 - 1 - 2 546 86

View Full Product Details