Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atxn7l2
Entrez Gene ID
72522
Gene Name
ataxin 7-like 2
Gene Aliases
2610528J18Rik
Chromosome Location
Chr.3: 108202222 - 108210527 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289545.1 NP_001276474.1 8 - 9 1391 66
NM_175183.5 NP_780392.1 9 - 10 1570 66
GenBank mRNA AK046849.1 - 9 - 10 1978 66
BC027136.1 - 5 - 6 1868 66
BC088996.1 - 7 - 8 1165 66
BC137599.1 - 8 - 9 1312 66
BC137600.1 - 8 - 9 1339 66
BC144756.1 - 8 - 9 1319 66

View Full Product Details