Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eogt
Entrez Gene ID
101351
Gene Name
EGF domain-specific O-linked N-acetylglucosamine (GlcNAc) transferase
Gene Aliases
A130022J15Rik, AI447490, AW214473, AW259391, Aer61
Chromosome Location
Chr.6: 97110016 - 97149703 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175313.4 NP_780522.1 16 - 17 1841 58
GenBank mRNA AK037507.1 - 16 - 17 1842 58
BC046414.1 - 9 - 10 842 58
BC048939.1 - 14 - 15 1380 58

View Full Product Details