Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eftud1
Entrez Gene ID
101592
Gene Name
elongation factor Tu GTP binding domain containing 1
Gene Aliases
4932434J20Rik, 6030468D11Rik, AI451340, AU019507, AU022896, D7Ertd791e
Chromosome Location
Chr.7: 82648614 - 82777852 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159672.1 NP_001153144.1 19 - 20 3250 63
NM_175317.3 NP_780526.2 19 - 20 3279 63
GenBank mRNA AK028676.1 - 19 - 20 3252 63
AK031647.1 - 19 - 20 3236 63
BC031852.1 - 13 - 14 2525 63
BC045616.1 - 19 - 20 3233 63

View Full Product Details