Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fcgbp
Entrez Gene ID
215384
Gene Name
Fc fragment of IgG binding protein
Gene Aliases
A430096B05Rik
Chromosome Location
Chr.7: 28071236 - 28120864 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK040441.1 - 1 - 1 329 60

View Full Product Details