The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040689.1 | NP_001035779.1 | 1 - 2 | 229 | 61 |
GenBank mRNA | AK020904.1 | - | 1 - 2 | 230 | 61 |
AK043484.1 | - | 1 - 2 | 230 | 61 | |
BC048707.1 | - | 1 - 2 | 203 | 61 | |
BC116367.1 | - | 1 - 2 | 192 | 61 |