The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175481.5 | NP_780690.2 | 10 - 11 | 1259 | 61 |
GenBank mRNA | AK032029.1 | - | 10 - 11 | 1260 | 61 |
AK080582.1 | - | 10 - 11 | 1387 | 61 | |
BC118010.2 | - | 9 - 10 | 1174 | 61 |