Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prtg
Entrez Gene ID
235472
Gene Name
protogenin homolog (Gallus gallus)
Gene Aliases
A230098A12Rik, Igdcc5
Chromosome Location
Chr.9: 72807274 - 72917307 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175485.4 NP_780694.3 3 - 4 710 67
GenBank mRNA AK083540.1 - 1 - 2 146 67
AY630257.1 - 3 - 4 780 67
BC116336.1 - 1 - 2 143 67
BC116337.1 - 1 - 2 143 67
DQ360114.1 - 3 - 4 710 67

View Full Product Details