The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175935.3 | NP_787949.2 | 1 - 2 | 444 | 73 |
GenBank mRNA | AK002966.1 | - | 2 - 3 | 308 | 73 |
AK165395.1 | - | 1 - 2 | 298 | 73 | |
AY186239.1 | - | 1 - 2 | 290 | 73 | |
BC069959.1 | - | 1 - 2 | 444 | 73 |