The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175937.3 | NP_787951.2 | 2 - 3 | 1911 | 118 |
GenBank mRNA | AB100307.1 | - | 3 - 4 | 544 | 118 |
BC107349.1 | - | 3 - 4 | 521 | 118 | |
BC107350.1 | - | 3 - 4 | 521 | 118 |