The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175937.3 | NP_787951.2 | 5 - 6 | 2141 | 84 |
GenBank mRNA | AB100307.1 | - | 6 - 7 | 774 | 84 |
BC107349.1 | - | 6 - 7 | 751 | 84 | |
BC107350.1 | - | 6 - 7 | 751 | 84 |