Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam76b
Entrez Gene ID
72826
Gene Name
family with sequence similarity 76, member B
Gene Aliases
2810485I05Rik, C78303
Chromosome Location
Chr.9: 13827702 - 13854687 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176836.3 NP_789806.1 8 - 9 1082 134
GenBank mRNA AK052430.1 - 8 - 9 1041 134
AK145026.1 - 8 - 9 1097 134
BC036328.1 - 1 - 2 276 134
BC043120.1 - 8 - 9 1082 134
BC050870.1 - 8 - 9 1000 134

View Full Product Details