Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Esrp2
Entrez Gene ID
77411
Gene Name
epithelial splicing regulatory protein 2
Gene Aliases
9530027K23Rik, Rbm35b
Chromosome Location
Chr.8: 106130403 - 106137528 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176838.2 NP_789808.1 3 - 4 649 75
GenBank mRNA AK088755.1 - 3 - 4 518 75
AK168488.1 - 3 - 4 497 75
BC031444.1 - 3 - 4 649 75
BC096581.1 - 3 - 4 489 75

View Full Product Details