The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177363.3 | NP_796337.1 | 5 - 6 | 812 | 84 |
GenBank mRNA | AK036904.1 | - | 5 - 6 | 813 | 84 |
AK089206.1 | - | - | 805 | 84 | |
AK155804.1 | - | 5 - 6 | 820 | 84 | |
AK170277.1 | - | 4 - 5 | 743 | 84 | |
DQ973493.1 | - | 5 - 6 | 764 | 84 |