Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ust
Entrez Gene ID
338362
Gene Name
uronyl-2-sulfotransferase
Gene Aliases
D930010O20Rik, UA2OST
Chromosome Location
Chr.10: 8204753 - 8518825 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177387.3 NP_796361.2 3 - 4 876 70
GenBank mRNA AK086181.1 - 3 - 4 823 70
AK147850.1 - 3 - 4 877 70
AK162950.1 - 3 - 4 449 70
BC138155.1 - 3 - 4 616 70

View Full Product Details