Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ofd1
Entrez Gene ID
237222
Gene Name
oral-facial-digital syndrome 1 gene homolog (human)
Gene Aliases
Cxorf5, DXGgc7e, ORF2
Chromosome Location
Chr.X: 166390033 - 166440838 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177429.3 NP_803178.2 17 - 18 2830 78
GenBank mRNA AJ278702.1 - 2 - 3 164 78
AJ438159.1 - 17 - 18 2681 78
AK141310.1 - 2 - 3 347 78
BC119070.1 - 17 - 18 2513 78
BC120487.1 - 17 - 18 2513 78

View Full Product Details