Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdc23
Entrez Gene ID
52563
Gene Name
CDC23 cell division cycle 23
Gene Aliases
6030435O18, D18Ertd243e
Chromosome Location
Chr.18: 34630946 - 34651745 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178347.4 NP_848124.1 15 - 16 1653 71
GenBank mRNA AK031459.1 - 15 - 16 1636 71
AK038523.1 - 15 - 16 1636 71
AK048246.1 - 15 - 16 1636 71
AK052567.1 - 15 - 16 1652 71
AK081461.1 - 15 - 16 1654 71
AK169677.1 - 11 - 12 1127 71
BC059013.1 - 15 - 16 1622 71

View Full Product Details