Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cidec
Entrez Gene ID
14311
Gene Name
cell death-inducing DFFA-like effector c
Gene Aliases
CIDE-3, Fsp27
Chromosome Location
Chr.6: 113424634 - 113435760 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301295.1 NP_001288224.1 2 - 3 122 66
NM_178373.3 NP_848460.1 2 - 3 106 66
GenBank mRNA AK080133.1 - 2 - 3 108 66
BC099676.1 - 1 - 2 78 66
M61737.1 - 2 - 3 122 66

View Full Product Details