The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178379.3 | NP_848466.1 | 6 - 7 | 1021 | 57 |
GenBank mRNA | AK010385.1 | - | 6 - 7 | 1004 | 57 |
AK149999.1 | - | 6 - 7 | 1011 | 57 | |
AK154668.1 | - | 6 - 7 | 1020 | 57 | |
AK171661.1 | - | 6 - 7 | 1011 | 57 | |
BC018301.1 | - | 6 - 7 | 972 | 57 | |
BC038046.1 | - | 6 - 7 | 974 | 57 |