Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h6
Entrez Gene ID
78751
Gene Name
zinc finger CCCH type containing 6
Gene Aliases
4631426G04Rik, 4833425H18Rik, AU024389, AW060453, Zc3hdc6, mKIAA2035
Chromosome Location
Chr.2: 128967402 - 129018563 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178404.3 NP_848491.2 8 - 9 1486 140
GenBank mRNA AK039171.1 - 8 - 9 1481 140
BC043311.1 - 4 - 5 442 140

View Full Product Details