Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krr1
Entrez Gene ID
52705
Gene Name
KRR1, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases
2610511F02Rik, AI255219, AI428520, D10Ertd773e, Hrb2
Chromosome Location
Chr.10: 111972695 - 111988430 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178610.4 NP_848725.2 1 - 2 181 63
GenBank mRNA AK012108.1 - 1 - 2 62
AK030560.1 - 1 - 2 93 63
AK032472.1 - 1 - 2 93 63
AK036447.1 - 1 - 2 102 63
AK041854.1 - 1 - 2 93 63
AK045526.1 - 1 - 2 120 63
AK146926.1 - 1 - 2 99 63
AK168675.1 - 1 - 2 93 63
BC089510.1 - 1 - 2 98 63
CD547185.1 - 1 - 2 181 63

View Full Product Details