The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164562.1 | NP_001158034.1 | 2 - 3 | 305 | 84 |
NM_178677.4 | NP_848792.2 | 3 - 4 | 430 | 84 | |
GenBank mRNA | AK029985.1 | - | 3 - 4 | 361 | 84 |
AK031125.1 | - | 2 - 3 | 302 | 84 | |
AK044249.1 | - | 2 - 3 | 305 | 84 | |
BB661643.1 | - | 3 - 4 | 430 | 84 | |
BC046289.1 | - | 2 - 3 | 260 | 84 |