The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178693.4 | NP_848808.1 | 3 - 4 | 320 | 76 |
GenBank mRNA | AK039422.1 | - | 3 - 4 | 321 | 76 |
AK039676.1 | - | 3 - 4 | 321 | 76 | |
AK043574.1 | - | 3 - 4 | 296 | 76 | |
AK050459.1 | - | 3 - 4 | 330 | 76 | |
AK163270.1 | - | - | 1089 | 76 | |
BC139373.1 | - | 3 - 4 | 363 | 76 | |
BC139376.1 | - | 3 - 4 | 360 | 76 | |
BC145615.1 | - | 3 - 4 | 363 | 76 |