The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098476.2 | NP_001091946.1 | 9 - 10 | 1201 | 128 |
NM_178700.5 | NP_848815.2 | 9 - 10 | 1529 | 128 | |
GenBank mRNA | AK045986.1 | - | 9 - 10 | 1483 | 128 |
AK077782.1 | - | 9 - 10 | 1530 | 128 | |
AK089014.1 | - | 8 - 9 | 1065 | 128 | |
AK168385.1 | - | 9 - 10 | 1173 | 128 | |
AK168459.1 | - | 9 - 10 | 1173 | 128 |