Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plscr4
Entrez Gene ID
235527
Gene Name
phospholipid scramblase 4
Gene Aliases
AV245873
Chromosome Location
Chr.9: 92457372 - 92492516 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178711.3 NP_848826.1 4 - 5 917 63
GenBank mRNA AK035878.1 - 4 - 5 902 63
AK035919.1 - 4 - 5 918 63
AK045372.1 - 4 - 5 918 63
AK054258.1 - 4 - 5 475 63
AK082330.1 - 3 - 4 360 63
AK141580.1 - - 443 63
AK165919.1 - 4 - 5 918 63
BC052067.1 - 4 - 5 471 63

View Full Product Details